Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007360.3 | NP_031386.2 | 8 | 739 | |
GenBank mRNA | AF260135.1 | - | 7 | 562 | |
AF260136.1 | - | 7 | 562 | ||
AK226161.1 | - | 6 | 1673 | ||
AK292059.1 | - | 8 | 725 | ||
BC039836.1 | - | 8 | 703 | 6067176 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199805.1 | NP_001186734.1 | 13 | 1097 | |
GenBank mRNA | AF439512.1 | - | 8 | 716 | |
AF461811.1 | - | 13 | 1106 | ||
X54870.1 | - | 10 | 900 |