Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007352.3 | NP_031378.1 | 7 | 846 | |
GenBank mRNA | AK092382.1 | - | 9 | 641 | |
AK315798.1 | - | 7 | 715 | ||
BC005216.1 | - | 7 | 720 | 3949903 | |
BM128949.1 | - | 2 | 198 | ||
M16630.1 | - | 7 | 691 | ||
M18692.1 | - | 7 | 718 |