Gene Transcripts

Gene Symbol
SNED1
Entrez Gene ID
25992
Gene Name
sushi, nidogen and EGF like domains 1
Gene Aliases
SST3, Snep
Chromosome Location
Chr. 2: 240997409 - 241095568 on Build GRCh38
UniGene ID
Hs.471834
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080437.1 NP_001073906.1 4 660
XM_011510931.2 XP_011509233.1 5 974
XM_011510932.2 XP_011509234.1 5 976
XM_011510933.2 XP_011509235.1 5 980
XM_011510934.2 XP_011509236.1 5 980
XM_011510937.2 XP_011509239.1 5 979
XM_017003773.1 XP_016859262.1 5 976
XM_017003774.1 XP_016859263.1 4 745
XM_017003775.1 XP_016859264.1 5 978
XM_017003776.1 XP_016859265.1 5 976
XM_017003777.1 XP_016859266.1 5 981
XM_017003778.1 XP_016859267.1 4 740
XM_017003779.1 XP_016859268.1 5 676
XM_017003780.1 XP_016859269.1 4 532
XM_017003781.1 XP_016859270.1 4 522
GenBank mRNA AK074062.1 - 3 268

View Full Product Details