Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001243399.1 | NP_001230328.1 | 4 | 863 | |
NM_001319291.1 | NP_001306220.1 | 4 | 444 | ||
NM_016066.4 | NP_057150.2 | 4 | 1021 | ||
NM_197962.2 | NP_932066.1 | 4 | 638 | ||
XM_017001397.1 | XP_016856886.1 | 4 | 571 | ||
GenBank mRNA | AA056513.1 | - | 1 | 112 | |
AA887118.1 | - | 1 | 115 | ||
AF132495.2 | - | 4 | 556 | ||
AF151891.1 | - | 4 | 528 | ||
AF290514.1 | - | 4 | 1021 | ||
AY038988.1 | - | 4 | 560 | ||
BC028113.1 | - | 4 | 544 | 5395272 | |
BM887926.1 | - | 4 | 444 | ||
BU157087.1 | - | 593 |