Gene Transcripts

Gene Symbol
POP5
Entrez Gene ID
51367
Gene Name
POP5 homolog, ribonuclease P/MRP subunit
Gene Aliases
HSPC004, RPP2, RPP20, hPop5
Chromosome Location
Chr. 12: 120579045 - 120581452 on Build GRCh38
UniGene ID
Hs.279913
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015918.3 NP_057002.2 5 459
NM_198202.1 NP_937845.1 4 313
XM_011538441.1 XP_011536743.1 5 571
GenBank mRNA AA918619.1 - 1 333
AF070660.1 - 5 443
AF117232.1 - 5 443
AJ306296.1 - 5 446
AK223206.1 - 5 431
AK303144.1 - 4 294
BC012505.1 - 5 446 4708484
BI668578.1 - 5 466
BU155588.1 - 302
CB995722.1 - 5 581

View Full Product Details