Gene Transcripts

Gene Symbol
BTBD7
Entrez Gene ID
55727
Gene Name
BTB domain containing 7
Gene Aliases
FUP1
Chromosome Location
Chr. 14: 93237550 - 93333092 on Build GRCh38
UniGene ID
Hs.525549
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002860.3 NP_001002860.2 3 1435
NM_018167.4 NP_060637.1 3 1435
XM_011536939.2 XP_011535241.1 3 1200
XM_017021438.1 XP_016876927.1 1 200
GenBank mRNA AB040958.2 - 3 1364
AK001510.1 - 3 1361
AK021953.1 - 1 654
AK303355.1 - 1 186
BC047071.1 - 3 1432
BC141850.1 - 3 1376

View Full Product Details