Gene Transcripts

Gene Symbol
LYRM2
Entrez Gene ID
57226
Gene Name
LYR motif containing 2
Gene Aliases
DJ122O8.2
Chromosome Location
Chr. 6: 89632224 - 89638755 on Build GRCh38
UniGene ID
Hs.177275
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020466.4 NP_065199.1 2 192
NR_028493.1 2 334
NR_028494.1 2 334
NR_028495.1 2 202
GenBank mRNA AK024783.1 - 2 190
AK094994.1 - 2 243
AK311151.1 - 2 334
AK311180.1 - 2 251
AK311275.1 - 2 187
AK311948.1 - 2 177
AL122090.1 - 2 175
AV710669.1 - 2 192
BC009782.1 - 2 186 4092710
BC104668.1 - 2 175 6717725
BP280644.1 - 2 192
DA160441.1 - 2 334

View Full Product Details