Gene Transcripts

Gene Symbol
LRFN1
Entrez Gene ID
57622
Gene Name
leucine rich repeat and fibronectin type III domain containing 1
Gene Aliases
SALM2
Chromosome Location
Chr. 19: 39306817 - 39320867 on Build GRCh38
UniGene ID
Hs.97860
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020862.1 NP_065913.1 1 162
XM_005259102.4 XP_005259159.1 4 362
XM_017027033.1 XP_016882522.1 4 477

View Full Product Details