Gene Transcripts

Gene Symbol
SFMBT2
Entrez Gene ID
57713
Gene Name
Scm-like with four mbt domains 2
Gene Aliases
Chromosome Location
Chr. 10: 7158624 - 7411486 on Build GRCh38
UniGene ID
Hs.407983
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018039.1 NP_001018049.1 4 314
NM_001029880.2 NP_001025051.1 4 416
XM_005252551.2 XP_005252608.1 4 388
XM_011519611.1 XP_011517913.1 4 415
XM_017016466.1 XP_016871955.1 4 317
GenBank mRNA AB046837.1 - 4 314
AK090887.1 - 4 422
BC144327.1 - 4 381
BC152430.1 - 4 314 8862546
DA152101.1 - 4 424

View Full Product Details