Gene Transcripts

Gene Symbol
MS4A6A
Entrez Gene ID
64231
Gene Name
membrane spanning 4-domains A6A
Gene Aliases
4SPAN3, 4SPAN3.2, CD20L3, CDA01, MS4A6, MST090, MSTP090
Chromosome Location
Chr. 11: 60171607 - 60184666 on Build GRCh38
UniGene ID
Hs.523702
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001247999.1 NP_001234928.1 6, 7 1117
NM_022349.3 NP_071744.2 5, 6 928
NM_152851.2 NP_690590.1 5, 6 824
NM_152852.2 NP_690591.1 6, 7 1053
XM_005274177.2 XP_005274234.1 6, 7 1112
XM_006718658.2 XP_006718721.1 6, 7 987
XM_006718660.2 XP_006718723.1 5, 6 852
XM_006718661.2 XP_006718724.1 5, 6 852
XM_017018123.1 XP_016873612.1 4, 5 588
XM_017018124.1 XP_016873613.1 4, 5 588
GenBank mRNA AB013104.1 - 5, 6 532
AF212240.1 - 5, 6 797
AF237908.1 - 5, 6 798
AF253977.1 - 5, 6 801
AF261136.1 - 5, 6 798
AF286866.1 - 4, 5 576
AF350502.1 - 5, 6 645
AF350503.1 - 5, 6 645
AF354930.1 - 5, 6 798
AK291870.1 - 6, 7 1053
AK301387.1 - 4, 5 488
BC108728.1 - 5, 6 701 6196213

View Full Product Details