Gene Transcripts

Gene Symbol
MOSPD3
Entrez Gene ID
64598
Gene Name
motile sperm domain containing 3
Gene Aliases
CDS3, NET30
Chromosome Location
Chr. 7: 100612102 - 100615380 on Build GRCh38
UniGene ID
Hs.521086
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040097.1 NP_001035186.1 4 590
NM_001040098.1 NP_001035187.1 4 506
NM_001040099.1 NP_001035188.1 3 659
NM_023948.4 NP_076438.1 3 689
XM_005250529.4 XP_005250586.1 4 511
XM_005250531.3 XP_005250588.1 4 536
GenBank mRNA BC005042.1 - 3 618
BC011653.2 - 3 670 4299899
BC028475.2 - 3 594
BQ923161.1 - 564

View Full Product Details