Gene Transcripts

Gene Symbol
CARD14
Entrez Gene ID
79092
Gene Name
caspase recruitment domain family member 14
Gene Aliases
BIMP2, CARMA2, PRP, PSORS2, PSS1
Chromosome Location
Chr. 17: 80169992 - 80209331 on Build GRCh38
UniGene ID
Hs.675480
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001257970.1 NP_001244899.1 5 872
NM_024110.4 NP_077015.2 5 872
NM_052819.2 NP_438170.1 2 153
NR_047566.1 6 937
XM_011525212.1 XP_011523514.1 7 1393
XM_011525213.1 XP_011523515.1 7 1135
XM_011525215.1 XP_011523517.1 6 1195
XM_011525216.1 XP_011523518.1 6 895
XM_011525217.1 XP_011523519.1 6 951
XM_011525218.2 XP_011523520.1 6 934
XM_017025057.1 XP_016880546.1 7 1393
XM_017025058.1 XP_016880547.1 7 1393
GenBank mRNA AF322642.1 - 5 910
AK091123.1 - 6 895
AK296762.1 - 2 93
AK309299.1 - 1 183
AX746790.1 - 6 895
AY032927.1 - 4 707
BC001326.1 - 2 153
BC018142.1 - 5 881 3847282
EU652409.1 - 4 704

View Full Product Details