Gene Transcripts
- Gene Symbol
- COQ5
- Entrez Gene ID
- 84274
- Gene Name
- coenzyme Q5, methyltransferase
- Gene Aliases
- Chromosome Location
- Chr. 12: 120503279 - 120529171 on Build GRCh38
- UniGene ID
- Hs.17250
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_032314.3 |
NP_115690.3
|
2, 3 |
382 |
|
XM_006719639.2 |
XP_006719702.1
|
3, 4 |
482 |
|
GenBank mRNA |
AK222610.1 |
- |
2, 3 |
372 |
|
AK225242.1 |
- |
2, 3 |
372 |
|
AK293656.1 |
- |
2, 3 |
403 |
|
AK315681.1 |
- |
2, 3 |
373 |
|
BC004916.2 |
- |
2, 3 |
358 |
|
BC107874.1 |
- |
2, 3 |
376 |
6452052 |
BP307378.1 |
- |
2, 3 |
382 |
|
BX647562.1 |
- |
2, 3 |
371 |
|
EU700459.1 |
- |
2, 3 |
376 |
|
View Full Product Details