Gene Transcripts
- Gene Symbol
- GOLGA2P2Y
- Entrez Gene ID
- 84559
- Gene Name
- golgin A2 pseudogene 2, Y-linked
- Gene Aliases
- GOLGA2LY, GOLGA2LY1, GOLGA2P2
- Chromosome Location
- Chr. Y: 24209967 - 24214831 on Build GRCh38
- UniGene ID
- Hs.531603
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001555.2 |
|
3 |
420 |
|
GenBank mRNA |
AF332229.1 |
- |
3 |
420 |
|
- Gene Symbol
- GOLGA2P3Y
- Entrez Gene ID
- 401634
- Gene Name
- golgin A2 pseudogene 3, Y-linked
- Gene Aliases
- GOLGA2LY2, GOLGA2P3
- Chromosome Location
- Chr. Y: 25455311 - 25460175 on Build GRCh38
- UniGene ID
- Hs.531603
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002195.2 |
|
3 |
420 |
|
View Full Product Details