Gene Transcripts

Gene Symbol
HES7
Entrez Gene ID
84667
Gene Name
hes family bHLH transcription factor 7
Gene Aliases
SCDO4, bHLHb37
Chromosome Location
Chr. 17: 8120590 - 8126032 on Build GRCh38
UniGene ID
Hs.434828
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001165967.1 NP_001159439.1 3 164
NM_032580.3 NP_115969.2 3 164
XM_011524041.2 XP_011522343.1 4 702
XM_017025232.1 XP_016880721.1 4 1758
GenBank mRNA AB049064.1 - 3 156
BC039887.1 - 3 162
BI259590.1 - 4 350

View Full Product Details