Gene Transcripts

Gene Symbol
KRTAP4-2
Entrez Gene ID
85291
Gene Name
keratin associated protein 4-2
Gene Aliases
KAP4.2, KRTAP4.2
Chromosome Location
Chr. 17: 41177446 - 41178208 on Build GRCh38
UniGene ID
Hs.380165
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033062.3 NP_149051.1 1 672
GenBank mRNA AJ406934.1 - 1 672

View Full Product Details