Gene Transcripts
- Gene Symbol
- TTTY3
- Entrez Gene ID
- 114760
- Gene Name
- testis-specific transcript, Y-linked 3 (non-protein coding)
- Gene Aliases
- LINC00121, NCRNA00121, TTTY3A, TTY3
- Chromosome Location
- Chr. Y: 25728490 - 25733388 on Build GRCh38
- UniGene ID
- Hs.511777
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001524.1 |
|
1 |
25 |
|
- Gene Symbol
- TTTY3B
- Entrez Gene ID
- 474148
- Gene Name
- testis-specific transcript, Y-linked 3B (non-protein coding)
- Gene Aliases
- LNCRNA00122, NCRNA00122
- Chromosome Location
- Chr. Y: 23936727 - 23941622 on Build GRCh38
- UniGene ID
- Hs.511777
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002176.1 |
|
1 |
25 |
|
View Full Product Details