Gene Transcripts
- Gene Symbol
- IFI27L1
- Entrez Gene ID
- 122509
- Gene Name
- interferon alpha inducible protein 27 like 1
- Gene Aliases
- FAM14B, ISG12C
- Chromosome Location
- Chr. 14: 94081293 - 94102714 on Build GRCh38
- UniGene ID
- Hs.19414
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145249.2 |
NP_660292.1
|
3, 4 |
303 |
|
NM_206949.2 |
NP_996832.1
|
3, 4 |
257 |
|
XM_005267314.4 |
XP_005267371.1
|
2, 3 |
2848 |
|
XM_011536405.2 |
XP_011534707.1
|
3, 4 |
273 |
|
XM_011536406.1 |
XP_011534708.1
|
4, 5 |
405 |
|
XM_011536407.1 |
XP_011534709.1
|
4, 5 |
408 |
|
XM_017020961.1 |
XP_016876450.1
|
3, 4 |
154 |
|
XM_017020963.1 |
XP_016876452.1
|
3, 4 |
156 |
|
GenBank mRNA |
BC015423.1 |
- |
3, 4 |
267 |
|
BU739372.1 |
- |
2, 3 |
426 |
|
View Full Product Details