Gene Transcripts

Gene Symbol
EARS2
Entrez Gene ID
124454
Gene Name
glutamyl-tRNA synthetase 2, mitochondrial
Gene Aliases
COXPD12, MSE1, gluRS
Chromosome Location
Chr. 16: 23522013 - 23557375 on Build GRCh38
UniGene ID
Hs.620541
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083614.1 NP_001077083.1 5 1027
NM_001308211.1 NP_001295140.1 5 1027
NR_003501.1 5 1027
XM_011545738.1 XP_011544040.1 5 1317
XM_011545739.1 XP_011544041.1 5 936
GenBank mRNA AB075850.1 - 5 984
AK095998.1 - 5 1016
AL832489.1 - 5 1003
BC040013.1 - 5 996 5728326
BX648129.1 - 3 706

View Full Product Details