Gene Transcripts
- Gene Symbol
- FAM53A
- Entrez Gene ID
- 152877
- Gene Name
- family with sequence similarity 53 member A
- Gene Aliases
- DNTNP
- Chromosome Location
- Chr. 4: 1574055 - 1684313 on Build GRCh38
- UniGene ID
- Hs.143314
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001013622.3 |
NP_001013644.1
|
2, 3 |
277 |
|
NM_001174070.2 |
NP_001167541.1
|
2, 3 |
324 |
|
NM_001297435.1 |
NP_001284364.1
|
2, 3 |
677 |
|
XM_005247948.3 |
XP_005248005.1
|
2, 3 |
974 |
|
XM_011513402.2 |
XP_011511704.1
|
2, 3 |
980 |
|
XM_011513403.1 |
XP_011511705.1
|
2, 3 |
974 |
|
XM_011513404.2 |
XP_011511706.1
|
2, 3 |
974 |
|
XM_011513405.1 |
XP_011511707.1
|
2, 3 |
974 |
|
XM_017007802.1 |
XP_016863291.1
|
2, 3 |
974 |
|
GenBank mRNA |
AK125566.1 |
- |
2, 3 |
272 |
|
BC070112.1 |
- |
2, 3 |
277 |
5268061 |
HY040719.1 |
- |
2, 3 |
330 |
|
View Full Product Details