Gene Transcripts
- Gene Symbol
- PRICKLE2
- Entrez Gene ID
- 166336
- Gene Name
- prickle planar cell polarity protein 2
- Gene Aliases
- EPM5
- Chromosome Location
- Chr. 3: 64093850 - 64445476 on Build GRCh38
- UniGene ID
- Hs.148105
- Species
- Homo sapiens
- Gene Symbol
- PRICKLE2-AS1
- Entrez Gene ID
- 100652759
- Gene Name
- PRICKLE2 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 64067964 - 64103131 on Build GRCh38
- UniGene ID
- Hs.731187
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045697.1 |
|
3 |
3224 |
|
GenBank mRNA |
AL832189.1 |
- |
3 |
3224 |
|
View Full Product Details