Gene Transcripts

Gene Symbol
CC2D1B
Entrez Gene ID
200014
Gene Name
coiled-coil and C2 domain containing 1B
Gene Aliases
Chromosome Location
Chr. 1: 52348437 - 52366291 on Build GRCh38
UniGene ID
Hs.591451
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032449.2 NP_115825.1 23 2577
XM_005270590.2 XP_005270647.1 23 2663
XM_005270591.2 XP_005270648.1 23 2645
XM_005270593.2 XP_005270650.1 19 2399
XM_006710423.2 XP_006710486.1 22 2537
XM_017000559.1 XP_016856048.1 23 2575
GenBank mRNA AB058739.1 - 19 2008
BC007912.2 - 11 1025
BC014080.2 - 19 2059
BC016880.1 - 17 1768 3913149
BC039308.1 - 21 3401

View Full Product Details