Gene Transcripts

Gene Symbol
ZBTB7C
Entrez Gene ID
201501
Gene Name
zinc finger and BTB domain containing 7C
Gene Aliases
APM-1, APM1, ZBTB36, ZNF857C
Chromosome Location
Chr. 18: 48026886 - 48412639 on Build GRCh38
UniGene ID
Hs.515388
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039360.2 NP_001034449.1 2 1381
NM_001318841.1 NP_001305770.1 4 1562
XM_005258229.4 XP_005258286.1 3 1306
XM_011525861.2 XP_011524163.1 6 1786
XM_011525863.2 XP_011524165.1 5 1903
XM_011525864.2 XP_011524166.1 5 1658
XM_011525865.2 XP_011524167.1 5 1899
XM_011525866.2 XP_011524168.1 4 1644
XM_011525867.2 XP_011524169.1 4 1433
XM_011525869.2 XP_011524171.1 5 1908
XM_011525870.2 XP_011524172.1 4 1604
XM_011525871.2 XP_011524173.1 2 2119
XM_017025605.1 XP_016881094.1 6 3581
XM_017025606.1 XP_016881095.1 6 2035
XM_017025607.1 XP_016881096.1 6 1790
XM_017025608.1 XP_016881097.1 5 3458
XM_017025609.1 XP_016881098.1 4 1775
GenBank mRNA AM988836.1 - 2 1369
AM988837.1 - 2 1514
AM988838.1 - 3 1564
AM988839.1 - 4 1562
EF120357.1 - 2 1433

View Full Product Details