Gene Transcripts

Gene Symbol
TMEM86B
Entrez Gene ID
255043
Gene Name
transmembrane protein 86B
Gene Aliases
Chromosome Location
Chr. 19: 55226634 - 55229264 on Build GRCh38
UniGene ID
Hs.135215
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173804.4 NP_776165.3 3 1058
XM_017026554.1 XP_016882043.1 2 831
GenBank mRNA AK126545.1 - 1 1458
BC023000.1 - 3 1058 5000011
BC033193.1 - 2 820

View Full Product Details