Gene Transcripts

Gene Symbol
MBLAC1
Entrez Gene ID
255374
Gene Name
metallo-beta-lactamase domain containing 1
Gene Aliases
Chromosome Location
Chr. 7: 100126694 - 100128498 on Build GRCh38
UniGene ID
Hs.632319
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203397.2 NP_981942.1 2 1009
XM_005250250.3 XP_005250307.1 2 1052
GenBank mRNA AW772344.1 - 1 483
BC031288.1 - 2 1006 5299411

View Full Product Details