Gene Transcripts

Gene Symbol
NAPSB
Entrez Gene ID
256236
Gene Name
napsin B aspartic peptidase, pseudogene
Gene Aliases
NAP1L, NAP2, NAPB, NAPSBP
Chromosome Location
Chr. 19: 50333800 - 50344748 on Build GRCh38
UniGene ID
Hs.636624
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002798.1 5 519
GenBank mRNA AF090387.1 - 5 526
AF098485.1 - 5 526
AF200344.1 - 5 527
AK300884.1 - 5 530
AK310134.1 - 5 537
BC085008.1 - 5 472
BC111705.1 - 5 516

View Full Product Details