Gene Transcripts

Gene Symbol
SIRPB2
Entrez Gene ID
284759
Gene Name
signal regulatory protein beta 2
Gene Aliases
PTPN1L, PTPNS1L3, dJ776F14.2
Chromosome Location
Chr. 20: 1474591 - 1491762 on Build GRCh38
UniGene ID
Hs.721685
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122962.1 NP_001116434.1 2 428
XM_005260708.4 XP_005260765.1 2 237
XM_011529221.2 XP_011527523.1 2 1024
XM_011529223.2 XP_011527525.1 2 603
XM_011529225.2 XP_011527527.1 1 63
GenBank mRNA AK095499.1 - 2 237
AK294440.1 - 2 254
BM285394.1 - 2 430
BQ635335.1 - 2 497
DC313096.1 - 2 326
DC358609.1 - 2 291

View Full Product Details