Gene Transcripts

Gene Symbol
BREA2
Entrez Gene ID
286076
Gene Name
breast cancer estrogen-induced apoptosis 2
Gene Aliases
Chromosome Location
Chr. 8: 143697115 - 143698413 on Build GRCh38
UniGene ID
Hs.178095
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015445.1 2 539
GenBank mRNA AL833948.1 - 2 539
AY940073.1 - 2 1508
BC132838.1 - 2 365
BC132840.1 - 2 365

View Full Product Details