Gene Transcripts

Gene Symbol
OR10A2
Entrez Gene ID
341276
Gene Name
olfactory receptor family 10 subfamily A member 2
Gene Aliases
OR10A2P, OR11-82, OR11-86, OST363
Chromosome Location
Chr. 11: 6869755 - 6870666 on Build GRCh38
UniGene ID
Hs.371762
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004460.1 NP_001004460.1 1 532
GenBank mRNA BC136955.1 - 1 594

View Full Product Details