Gene Transcripts

Gene Symbol
MURC
Entrez Gene ID
347273
Gene Name
muscle related coiled-coil protein
Gene Aliases
CAVIN4, cavin-4
Chromosome Location
Chr. 9: 100578079 - 100588389 on Build GRCh38
UniGene ID
Hs.733374
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018116.2 NP_001018126.1 1 275
GenBank mRNA AK300099.1 - 1 275
BC090888.1 - 1 192 30529833
EU487253.1 - 1 210

View Full Product Details