Gene Transcripts

Gene Symbol
GLYATL3
Entrez Gene ID
389396
Gene Name
glycine-N-acyltransferase like 3
Gene Aliases
C6orf140, bA28H17.2
Chromosome Location
Chr. 6: 49499958 - 49528064 on Build GRCh38
UniGene ID
Hs.585172
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010904.1 NP_001010904.1 2 129
GenBank mRNA DB246049.1 - 2 129

View Full Product Details