Gene Transcripts

Gene Symbol
RP1
Entrez Gene ID
6101
Gene Name
retinitis pigmentosa 1 (autosomal dominant)
Gene Aliases
DCDC4A, ORP1
Chromosome Location
Chr. 8: 54509448 - 54630834 on Build GRCh38
UniGene ID
Hs.732820
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK098176.1 - 9 1161
AK302348.1 - 6 761
AK304443.1 - 8 922
AK316511.1 - 8 1356

Gene Symbol
LOC107984125
Entrez Gene ID
107984125
Gene Name
lipoxygenase homology domain-containing protein 1-like
Gene Aliases
Chromosome Location
Chr. 8: 54657854 - 54871720 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017014157.1 XP_016869646.1 2 370

View Full Product Details