Gene Transcripts

Gene Symbol
SH2D5
Entrez Gene ID
400745
Gene Name
SH2 domain containing 5
Gene Aliases
Chromosome Location
Chr. 1: 20719731 - 20732640 on Build GRCh38
UniGene ID
Hs.591522
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001103160.1 NP_001096630.1 5 928
NM_001103161.1 NP_001096631.1 6 1057
XM_011541459.1 XP_011539761.1 6 1522
XM_011541460.1 XP_011539762.1 7 1362
XM_011541461.2 XP_011539763.1 6 635
XM_011541462.1 XP_011539764.1 6 1522

View Full Product Details