Gene Transcripts

Gene Symbol
KRTAP5-6
Entrez Gene ID
440023
Gene Name
keratin associated protein 5-6
Gene Aliases
KRTAP5.6
Chromosome Location
Chr. 11: 1697195 - 1697755 on Build GRCh38
UniGene ID
Hs.534863
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012416.1 NP_001012416.1 1 497
GenBank mRNA AB126075.1 - 1 497
BC130399.1 - 1 469 40146430
BC133035.1 - 1 469 40147057

View Full Product Details