Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001277783.1 | NP_001264712.1 | 3 | 3338 | |
NM_004707.3 | NP_004698.3 | 4 | 3475 | ||
NR_033362.1 | 5 | 3592 | |||
NR_033363.1 | 5 | 3591 | |||
NR_073603.1 | 6 | 3708 | |||
GenBank mRNA | AB002449.2 | - | 1 | 312 |