Gene Transcripts
- Gene Symbol
- ERICH1
- Entrez Gene ID
- 157697
- Gene Name
- glutamate rich 1
- Gene Aliases
- HSPC319
- Chromosome Location
- Chr. 8: 614737 - 731239 on Build GRCh38
- UniGene ID
- Hs.655310
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK090404.1 |
- |
2 |
2756 |
|
View Full Product Details