Gene Transcripts
- Gene Symbol
- RAB1B
- Entrez Gene ID
- 81876
- Gene Name
- RAB1B, member RAS oncogene family
- Gene Aliases
- Chromosome Location
- Chr. 11: 66268526 - 66277492 on Build GRCh38
- UniGene ID
- Hs.300816
- Species
- Homo sapiens
- Gene Symbol
- RAB1C
- Entrez Gene ID
- 441400
- Gene Name
- RAB1C, member RAS oncogene family pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 9: 37636688 - 37637293 on Build GRCh38
- UniGene ID
- Hs.651788
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U66622.1 |
- |
1 |
145 |
|
View Full Product Details