Gene Transcripts

Gene Symbol
IMMP2L
Entrez Gene ID
83943
Gene Name
inner mitochondrial membrane peptidase subunit 2
Gene Aliases
IMMP2L-IT1, IMP2, IMP2-LIKE
Chromosome Location
Chr. 7: 110662645 - 111562531 on Build GRCh38
UniGene ID
Hs.655722
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011516608.2 XP_011514910.1 8 1158
XM_017012704.1 XP_016868193.1 7 2268

View Full Product Details