Gene Transcripts
- Gene Symbol
- SPANXD
- Entrez Gene ID
- 64648
- Gene Name
- SPANX family member D
- Gene Aliases
- CT11.4, SPANX-C, SPANX-D, SPANX-E, SPANXE, dJ171K16.1
- Chromosome Location
- Chr. X: 141697401 - 141698739 on Build GRCh38
- UniGene ID
- Hs.375036
- Species
- Homo sapiens
- Gene Symbol
- SPANXB1
- Entrez Gene ID
- 728695
- Gene Name
- SPANX family member B1
- Gene Aliases
- B1, CT11.2, SPANX-B, SPANXB, SPANXB2, SPANXF1, SPANXF2
- Chromosome Location
- Chr. X: 141002591 - 141003706 on Build GRCh38
- UniGene ID
- Hs.434105
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF098307.1 |
- |
2 |
313 |
|
BC034472.1 |
- |
2 |
311 |
4824918 |
- Gene Symbol
- SPANXC
- Entrez Gene ID
- 64663
- Gene Name
- SPANX family member C
- Gene Aliases
- C, CT11.3, CTp11, SPANX-C
- Chromosome Location
- Chr. X: 141241463 - 141242550 on Build GRCh38
- UniGene ID
- Hs.375036
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022661.3 |
NP_073152.2
|
2 |
285 |
|
View Full Product Details