Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136504.1 | NP_001129976.1 | 9 | 6214 | |
NM_177402.4 | NP_796376.2 | 9 | 6204 | ||
XM_011509192.2 | XP_011507494.1 | 9 | 6234 | ||
XM_017000309.1 | XP_016855798.1 | 8 | 7980 | ||
XM_017000310.1 | XP_016855799.1 | 8 | 7971 | ||
XM_017000311.1 | XP_016855800.1 | 9 | 6223 | ||
XM_017000312.1 | XP_016855801.1 | 9 | 6888 | ||
XM_017000313.1 | XP_016855802.1 | 9 | 6878 | ||
GenBank mRNA | AK124510.1 | - | 1 | 2525 |