Gene Transcripts

Gene Symbol
ELFN2
Entrez Gene ID
114794
Gene Name
extracellular leucine rich repeat and fibronectin type III domain containing 2
Gene Aliases
LRRC62, PPP1R29
Chromosome Location
Chr. 22: 37340644 - 37427470 on Build GRCh38
UniGene ID
Hs.344170
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052906.4 NP_443138.2 3 2001
GenBank mRNA AB067491.1 - 1 1220

View Full Product Details