Gene Transcripts

Gene Symbol
ASB12
Entrez Gene ID
142689
Gene Name
ankyrin repeat and SOCS box containing 12
Gene Aliases
Chromosome Location
Chr. X: 64224192 - 64230631 on Build GRCh38
UniGene ID
Hs.56281
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130388.3 NP_569059.3 2 681
GenBank mRNA AF403030.1 - 1 461
AK096896.1 - 2 661
AK300684.1 - 2 657
BC069436.1 - 1 461 7262244
BC069555.1 - 1 461 7262256
BC093883.1 - 1 522 7939728
BC104946.1 - 1 522 8143949

View Full Product Details