Gene Transcripts

Gene Symbol
OTOA
Entrez Gene ID
146183
Gene Name
otoancorin
Gene Aliases
CT108, DFNB22
Chromosome Location
Chr. 16: 21663857 - 21771468 on Build GRCh38
UniGene ID
Hs.408336
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161683.1 NP_001155155.1 14 1809
NM_144672.3 NP_653273.3 18 1953
NM_170664.2 NP_733764.1 9 1047
XM_011545747.2 XP_011544049.1 19 2332
XM_011545748.2 XP_011544050.2 8 1326
XM_017022950.1 XP_016878439.1 19 2056
XM_017022951.1 XP_016878440.1 4 350
GenBank mRNA AK093062.1 - 9 1047
AK125840.1 - 14 1809
AK302733.1 - 8 1109
BC038458.1 - 2 179
BC129992.1 - 18 1950
BC129993.1 - 18 1957 40128985

View Full Product Details