Gene Transcripts

Gene Symbol
DHODH
Entrez Gene ID
1723
Gene Name
dihydroorotate dehydrogenase (quinone)
Gene Aliases
DHOdehase, POADS, URA1
Chromosome Location
Chr. 16: 72008744 - 72027659 on Build GRCh38
UniGene ID
Hs.654427
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001361.4 NP_001352.2 2, 3 266
XM_005255827.3 XP_005255884.1 2, 3 571
XM_005255829.3 XP_005255886.1 1 33
XM_017022990.1 XP_016878479.1 2, 3 346
GenBank mRNA AK292293.1 - 3, 4 422
AU130529.1 - 3, 4 266
BC065245.1 - 2, 3 246 6064723
BI253747.1 - 2, 3 250
BM795077.1 - 1, 2 41
M94065.1 - 3, 4 248

View Full Product Details