Gene Transcripts

Gene Symbol
OR6Q1
Entrez Gene ID
219952
Gene Name
olfactory receptor family 6 subfamily Q member 1 (gene/pseudogene)
Gene Aliases
OR11-226
Chromosome Location
Chr. 11: 58030953 - 58031906 on Build GRCh38
UniGene ID
Hs.690432
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005186.2 NP_001005186.2 1 172
GenBank mRNA BC151146.1 - 1 195
BC151147.1 - 1 195 9021018

View Full Product Details