Gene Transcripts

Gene Symbol
CCDC89
Entrez Gene ID
220388
Gene Name
coiled-coil domain containing 89
Gene Aliases
Chromosome Location
Chr. 11: 85683844 - 85686277 on Build GRCh38
UniGene ID
Hs.376241
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152723.2 NP_689936.1 1 642
GenBank mRNA AK095478.1 - 1 642
BC044814.1 - 1 538 5167843
BI559499.1 - 1 598

View Full Product Details