Gene Transcripts
- Gene Symbol
- VASH1
- Entrez Gene ID
- 22846
- Gene Name
- vasohibin 1
- Gene Aliases
- KIAA1036
- Chromosome Location
- Chr. 14: 76761457 - 76783020 on Build GRCh38
- UniGene ID
- Hs.525479
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014909.4 |
NP_055724.1
|
6, 7 |
1964 |
|
XM_017021087.1 |
XP_016876576.1
|
8, 9 |
1039 |
|
XM_017021088.1 |
XP_016876577.1
|
7, 8 |
996 |
|
XM_017021089.1 |
XP_016876578.1
|
7, 8 |
855 |
|
XM_017021090.1 |
XP_016876579.1
|
6, 7 |
788 |
|
XM_017021091.1 |
XP_016876580.1
|
6, 7 |
2464 |
|
GenBank mRNA |
AB028959.1 |
- |
6, 7 |
1419 |
|
AK291377.1 |
- |
6, 7 |
1114 |
|
AL832588.2 |
- |
6, 7 |
1668 |
|
BC051896.1 |
- |
6, 7 |
1258 |
5170122 |
View Full Product Details