Gene Transcripts

Gene Symbol
TNRC6B
Entrez Gene ID
23112
Gene Name
trinucleotide repeat containing 6B
Gene Aliases
Chromosome Location
Chr. 22: 40044817 - 40335808 on Build GRCh38
UniGene ID
Hs.372082
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024843.1 NP_001020014.1 20 2533
NM_001162501.1 NP_001155973.1 19 4854
NM_015088.2 NP_055903.2 17 4524
GenBank mRNA AB029016.2 - 17 4504
AK294519.1 - 19 4845
AK304087.1 - 14 2009
BC028626.2 - 20 2466 4826787
BX647630.1 - 14 3726

View Full Product Details