Gene Transcripts

Gene Symbol
FBXL7
Entrez Gene ID
23194
Gene Name
F-box and leucine rich repeat protein 7
Gene Aliases
FBL6, FBL7
Chromosome Location
Chr. 5: 15500196 - 15939796 on Build GRCh38
UniGene ID
Hs.433057
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278317.1 NP_001265246.1 2 334
NM_012304.4 NP_036436.1 2 566
XM_011513998.1 XP_011512300.1 1 48
XM_017009262.1 XP_016864751.1 2 4546
GenBank mRNA AB020647.2 - 2 566
AF174593.1 - 1 61
AF199356.1 - 2 89
AK309585.1 - 2 334
BC075061.2 - 2 134 30915585
BC136424.1 - 2 331 9020413

View Full Product Details