Gene Transcripts

Gene Symbol
ARC
Entrez Gene ID
23237
Gene Name
activity regulated cytoskeleton associated protein
Gene Aliases
Arg3.1
Chromosome Location
Chr. 8: 142611044 - 142614472 on Build GRCh38
UniGene ID
Hs.40888
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015193.4 NP_056008.1 1 1194
GenBank mRNA AF193421.1 - 1 1182
BC012321.1 - 1 1198 4555433
BC075802.1 - 1 1186 5198843
D87468.1 - 1 1186

View Full Product Details